Skip to Content
Merck
All Photos(1)

Documents

54040-U

Supelco

SPE tube

Si/44%H2SO4/Si SPE Tube, volume 12mL, pk of 20 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

product name

12mL Si/44%H2SO4/Si SPE Tube, size 12 mL, 0.2 g (Silica), 3 g (44% H2SO4 Silica), 0.2 g (Silica), pk of 20 ea

composition

, 0.2 g (Silica)
, 3 g (44% H2SO4 Silica)

packaging

pk of 20 ea

technique(s)

solid phase extraction (SPE): suitable

size

12 mL

application(s)

food and beverages

storage temp.

2-8°C

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuki Haga et al.
Environmental science and pollution research international, 25(17), 16446-16454 (2018-04-16)
In this study, we analyzed hydroxylated polychlorinated biphenyls (OH-PCBs) in urine of both PCB transport workers and PCB researchers. A method to monitor OH-PCB in urine was developed. Urine was solid-phase extracted with 0.1% ammonia/ methanol (v/v) and glucuronic acid/sulfate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service