Skip to Content
Merck
All Photos(1)

Key Documents

52751-U

Supelco

Discovery® DSC-Diol SPE Tube

bed wt. 500 mg, volume 3 mL, pk of 54

Sign Into View Organizational & Contract Pricing

Select a Size

54 EA
€277.00

€277.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest a Sample

Select a Size

Change View
54 EA
€277.00

About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

€277.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest a Sample

material

polypropylene tube

composition

bed wt., 500 mg

packaging

pk of 54

extent of labeling

7% C loading

technique(s)

solid phase extraction (SPE): suitable

surface area

480 m2/g

volume

3 mL

matrix

Silica gel base material (irregularly shaped, acid washed)

matrix active group

diol bonding, polymerically bonded

particle size

50 μm

pore size

0.9 mL/g pore volume
70 Å pore diameter

application(s)

food and beverages

separation technique

normal phase

Looking for similar products? Visit Product Comparison Guide

General description

Retention Mechanism: Normal phase
Sample Matrix Compatibility: Organic solvents, oils, and lipids

  • Polymerically bonded, 2,3-Dihydroxypropoxypropyl (7% C)
  • Polar sorbent most commonly used for normal phase applications (polar extractions from non-polar matrices)
  • The sorbent dihydroxy groups facilitate strong hydrogen bonding
  • Excellent selectivity when extracting structurally similar molecules

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 2 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service