50645-U
Ascentis® Express 90 Å PCS-C18, 2.7 µm HPLC Column
column L × I.D. 150 mm × 3 mm, pk of 1 ea
All Photos(1)
About This Item
Recommended Products
General description
Ascentis® Express PCS-C18 columns are specifically engineered to provide efficient separation of basic, acidic, or neutral compounds under low ionic strength (formic acid) mobile phase conditions. These columns contain advanced Fused-Core® particles with pores strategically sized to 90 Å for small molecule analyses. This attribute makes these columns an excellent choice for pharmaceutical method development. The unique PCS-C18 chemistry enables exceptional peak shape and enhanced loading capacity of basic compounds as compared to traditional C18 chemistries. This column also provides orthogonal selectivity to other small molecule chemistries.
Analysis Note
The target analytes include basic compounds.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
Fused-Core is a registered trademark of Advanced Materials Technology, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)