Skip to Content
Merck
All Photos(1)

Key Documents

505013-30

Supelco

Discovery® RP-Amide C16 (5 µm) HPLC Columns

L × I.D. 15 cm × 3 mm, HPLC Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€801.00

€801.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€801.00

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

€801.00


Please contact Customer Service for Availability

Product Name

Discovery® RP-AmideC16 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 3 mm

material

stainless steel column

Agency

suitable for USP L60

product line

Discovery®

feature

endcapped

manufacturer/tradename

Discovery®

packaging

1 ea of

extent of labeling

11% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

15 cm × 3 mm

surface area

200 m2/g

surface coverage

2.6 μmol/m2

impurities

<10 ppm metals

matrix

silica gel, high purity, spherical base material
fully porous particle

matrix active group

amide, alkyl phase

particle size

5 μm

pore size

180 Å

operating pH range

2-8

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Excellent retention and resolution of polar compounds
  • Unique selectivity compared to C18
  • Excellent peak shape and efficiency
  • Less hydrophobic than C18 columns
  • Compatible with 100% aqueous mobile phases
   

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annamaria Jakab et al.
Journal of pharmaceutical and biomedical analysis, 27(6), 913-921 (2002-02-12)
Retention parameters of 45 different barbituric acid derivatives were determined on an amide embedded RP silica column (Discovery RP-AmideC16) using non-buffered water-acetonitrile eluent systems. Linear correlation were calculated between the logarithm of the capacity factor and the acetonitrile concentration in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service