Skip to Content
Merck
All Photos(1)

Key Documents

50136-U

Supelco

LiChrospher® RP-18 (5 µm) HPLC Columns

L × I.D. 12.5 cm × 4.6 mm, HPLC Column

Synonym(s):

TM=["LiChrospher"] C18 HPLC column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€768.00

€768.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€768.00

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

€768.00


Please contact Customer Service for Availability

Product Name

LiChrospher® RP-18 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4.6 mm

material

stainless steel column

Agency

suitable for USP L1

product line

LiChrospher®

feature

endcapped: no

manufacturer/tradename

LiChrospher®

packaging

1 ea of

extent of labeling

21% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

12.5 cm × 4.6 mm

surface area

350 m2/g

matrix

octadecyl
silica particle platform
fully porous particle

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

100 Å

operating pH

2-7.5

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

LiChrospher® RP-18 HPLC column (L × I.D. 12.5 cm × 4.6 mm) is generally found to be highly reliable and versatile. It is suitable for reversed-phase high performance liquid chromatography (RP-HPLC) of acidic, neutral and weakly basic compounds, according to L1 USP specifications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChrospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service