Skip to Content
Merck
All Photos(1)

Key Documents

49476-U

Supelco

Benzo(k)fluoranthene solution

100 μg/mL in methylene chloride, analytical standard

Synonym(s):

Benzo[k]fluoranthene solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
Beilstein:
1873745
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

availability

available only in Italy

concentration

100 μg/mL in methylene chloride

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

SMILES string

c1ccc2cc-3c(cc2c1)-c4cccc5cccc-3c45

InChI

1S/C20H12/c1-2-6-15-12-19-17-10-4-8-13-7-3-9-16(20(13)17)18(19)11-14(15)5-1/h1-12H

InChI key

HAXBIWFMXWRORI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzo(k)fluoranthene belongs to the class of polynuclear aromatic hydrocarbons (PAHs). PAHs are a group of well-known environmental pollutants with both mutagenic and carcinogenic properties. They are released into the environment by the pyrolysis of organic materials.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

E J LaVoie et al.
Cancer research, 40(12), 4528-4532 (1980-12-01)
The metabolism of the environmental agents benzo(j)-fluoranthene and benzo(k)fluoranthene was investigated using supernatants from the livers of Aroclor 1254-pretreated rats, which are effective in activating benzo(j)fluoranthene and benzo(k)fluoranthene to metabolites mutagenic toward Salmonella typhimurium TA 100. Six bands of metabolites
Erin E Bessette et al.
Drug metabolism and disposition: the biological fate of chemicals, 33(3), 312-320 (2004-12-04)
Polycyclic aromatic hydrocarbons (PAHs) and heavy metals are often environmental cocontaminants that could interact to alter PAH carcinogenicity. The heavy metal, arsenite, and the PAH, benzo[k]fluoranthene, were used as prototypes to investigate, in human HepG2 cells, mechanisms whereby the bioactivation
Hiroyuki Masaki et al.
Environmental sciences : an international journal of environmental physiology and toxicology, 13(6), 353-362 (2007-02-03)
We developed a specific analysis method for benzo[a]pyrene (B[a]P) and benzo[k]fluoranthene (B[k]F) using dual beam fluorometry. B[a]P and B[k]F exhibited two specific fluorescence intensities (up to 10-fold) compared with 22 other polycyclic aromatic hydrocarbons (PAHs) at 370 nm excitation and
Tohru Saitoh et al.
Talanta, 79(2), 177-182 (2009-06-30)
A simple and rapid method for the highly sensitive determination of polycyclic aromatic hydrocarbons (PAHs) in water was developed. Benzo[a]pyrene, benzo[k]fluoranthene, perylene, and pyrene in water were concentrated into sodium dodecyl sulfate (SDS)-alumina admicelles. The collection was performed by adding
G Verrhiest et al.
Ecotoxicology (London, England), 10(6), 363-372 (2002-01-05)
Polycyclic Aromatic Hydrocarbons (PAHs) are ubiquitous pollutants of sediments. Sediment quality criteria often use toxicity data for individual PAHs. However, PAHs always occur in field sediments as a complex mixture of compounds. In this study, the toxicity of phenanthrene (P)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service