Skip to Content
Merck
All Photos(1)

Key Documents

48924-U

Supelco

Pre-Activated Florisil®

ampulized, for use with Dioxin Sample Preparation Kit, 60-100 mesh particle size, pk of 10

Synonym(s):

activated Florisil/magnesium silicate

Sign Into View Organizational & Contract Pricing

Select a Size

10 X 1 G
€82.40
1 G
€130.00
10 X 1.5 G
€145.00

€82.40


Please contact Customer Service for Availability

Request a Bulk OrderRequest a Sample

Select a Size

Change View
10 X 1 G
€82.40
1 G
€130.00
10 X 1.5 G
€145.00

About This Item

EC Number:
UNSPSC Code:
13111000
NACRES:
NB.21

€82.40


Please contact Customer Service for Availability

Request a Bulk OrderRequest a Sample

Agency

suitable for EPA 1613

Quality Level

packaging

pk of 10

technique(s)

solid phase extraction (SPE): suitable

particle size

60-100 mesh

application(s)

food and beverages

compatibility

for use with Dioxin Sample Preparation Kit

General description

Pre-activated ampulized Florisil is intended for use with the Supelco Dioxin Prep System to affect the class separation of polychlorinated biphenyls (PCBs) from polychlorinated dibenzo-p-dioxins and dibenzo-furans (PCDD/Fs).

Analysis Note

How to Use the Pre-activated Florisil®:
  1. Insert glass wool plug into the tapered end of a graduated serological pipette
  2. Snap the ampul and empty contents completely into the serological pipette
  3. Top the Florisil® with approximately 1 mL of sodium sulfate and a glass wool plug
  4. Carefully dispose of the empty glass ampul
  5. Condition the bed to remove pockets of air and to wet the surface ofthe material using appropriate solvent or solvent combination
  6. Follow the remaining steps in EPA method 1613b.

Legal Information

Florisil is a registered trademark of U.S. Silica Company

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service