Skip to Content
Merck
All Photos(1)

Key Documents

48715-U

Supelco

Naphthalene-d8 solution

certified reference material, 2000 μg/mL in dichloromethane

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€47.90

€47.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€47.90

About This Item

CAS Number:
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24

€47.90


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 625

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type internal/surrogate

packaging

ampule of 1 mL

concentration

2000 μg/mL in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

SMILES string

c21c(c(c(c(c2[2H])[2H])[2H])[2H])c(c(c(c1[2H])[2H])[2H])[2H]

InChI

1S/C10H8/c1-2-6-10-8-4-3-7-9(10)5-1/h1-8H/i1D,2D,3D,4D,5D,6D,7D,8D

InChI key

UFWIBTONFRDIAS-PGRXLJNUSA-N

General description

Naphthalene-d8 solution is an analytical reference standard specifically designed for monitoring organic chemicals on the Priority Pollutants List in municipal and industrial wastewater per methods developed by the US EPA Environmental Monitoring Systems Laboratory in Cincinnati, Ohio (EMSL/CL), under authority of the Clean Water Act (CWA).

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support. Naphthalene-d8 solution is used as an internal standard for the analysis of polynuclear aromatic hydrocarbons (PAHs) in olive oil using a novel SPE cartridge followed by gas chromatography-mass spectrometric (GC-MS) analysis.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT RE 2 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service