Skip to Content
Merck
All Photos(1)

Key Documents

48505

Supelco

Carbon tetrachloride

analytical standard

Synonym(s):

Tetrachloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CCl4
CAS Number:
Molecular Weight:
153.82
Beilstein:
1098295
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

5.32 (vs air)

vapor pressure

143 mmHg ( 30 °C)
91 mmHg ( 20 °C)

packaging

ampule of 1000 mg

availability

not available in Japan

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.460 (lit.)

bp

76-77 °C (lit.)

mp

−23 °C (lit.)

density

1.594 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

ClC(Cl)(Cl)Cl

InChI

1S/CCl4/c2-1(3,4)5

InChI key

VZGDMQKNWNREIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Ozone 1 - Skin Sens. 1B - STOT RE 1 Inhalation

Target Organs

Liver,Kidney

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samar Basu
Toxicology, 189(1-2), 113-127 (2003-06-25)
Hepatic injury through carbon tetrachloride (CCl(4)) induced lipid peroxidation is well known and has been extensively used in the experimental models to understand the cellular mechanisms behind oxidative damage and further to evaluate the therapeutic potential of drugs and dietary
Bi-Sen Ding et al.
Nature, 505(7481), 97-102 (2013-11-22)
Chemical or traumatic damage to the liver is frequently associated with aberrant healing (fibrosis) that overrides liver regeneration. The mechanism by which hepatic niche cells differentially modulate regeneration and fibrosis during liver repair remains to be defined. Hepatic vascular niche
Xingbin Yang et al.
Food chemistry, 138(2-3), 1256-1264 (2013-02-16)
Water-soluble apple peel polysaccharides (APP) and apple flesh polysaccharides (AFP) were isolated from Pink Lady fruits, and their in vitro antioxidant capacities were characterised by DPPH(), HO(), and O(2)(-) systems, and ferric-reducing antioxidant power assay. Oral administration of APP at
Marc Mejias et al.
Gut, 64(4), 657-666 (2014-05-23)
Antiangiogenic strategies have been proposed as a promising new approach for the therapy of portal hypertension and chronic liver disease. Pigment epithelium-derived factor (PEDF) is a powerful endogenous angiogenesis inhibitor whose role in portal hypertension remains unknown. Therefore, we aimed
Inge Mannaerts et al.
PloS one, 8(1), e55786-e55786 (2013-02-06)
The conversion of a quiescent vitamin A storing hepatic stellate cell (HSC) to a matrix producing, contractile myofibroblast-like activated HSC is a key event in the onset of liver disease following injury of any aetiology. Previous studies have shown that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service