Skip to Content
Merck
All Photos(1)

Documents

48318

Supelco

Decachlorobiphenyl solution

certified reference material, 200 μg/mL in acetone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12Cl10
CAS Number:
Molecular Weight:
498.66
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

certified reference material
TraceCERT®

Agency

EPA 8082,8310

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type surrogate

packaging

ampule of 1 mL

concentration

200 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

SMILES string

Clc1c(Cl)c(Cl)c(c(Cl)c1Cl)-c2c(Cl)c(Cl)c(Cl)c(Cl)c2Cl

InChI

1S/C12Cl10/c13-3-1(4(14)8(18)11(21)7(3)17)2-5(15)9(19)12(22)10(20)6(2)16

InChI key

ONXPZLFXDMAPRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Photodegradation of decachlorobiphenyl.
M Koshioka et al.
Bulletin of environmental contamination and toxicology, 38(3), 409-415 (1987-03-01)
A morphologic and physiologic bioassay for quantifying alterations in the lung following experimental chronic inhalation of tobacco smoke.
G L Huber et al.
Bulletin europeen de physiopathologie respiratoire, 17(2), 269-327 (1981-01-01)
Perchlorination reaction applied to the rapid determination of PCBs.
K Takamiya
Bulletin of environmental contamination and toxicology, 30(5), 600-605 (1983-05-01)
Use of two halogenated biphenyls as indicators of non-target exposure during rodenticide treatments.
M G Townsend et al.
Bulletin of environmental contamination and toxicology, 54(4), 526-533 (1995-04-01)
Xing Han et al.
Toxicology, 255(3), 177-186 (2008-11-22)
2,2',3,3',4,4',5,5',6,6'-Decachlorobiphenyl (PCB 209) is a fully chlorinated, non-coplanar biphenyl. To demonstrate that PCB 209 is not likely to exhibit human health hazards common to coplanar PCBs it was tested for cytochrome P450 (P450) enzyme induction potentials, genetic toxicity, and endocrine-modulating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service