Skip to Content
Merck
All Photos(1)

Key Documents

48055-U

Supelco

Aroclor 1254 solution

certified reference material, 1000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

InChI

1S/C12H5Cl5/c13-8-3-1-2-6(10(8)15)7-4-5-9(14)12(17)11(7)16/h1-5H

InChI key

AUGNBQPSMWGAJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aroclor 1254 is found to be a complex mixture of polychlorinated biphenyl (PCB) congeners and is prepared by the chlorination of biphenyl.[1]

Application

Aroclor 1248 may be used as a certified reference standard for the quantification of polychlorinated biphenyls (PCBs) in environmental matrices[1] and transformer oil samples[2] using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 1 - Flam. Liq. 2 - STOT RE 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Levels of polychlorinated biphenyls (PCBs) in transformer oils from Korea
Shin KS, et al.
Journal of Hazardous Materials, 137(3), 1514-1522 (2006)
Jing An et al.
Toxicology, 295(1-3), 8-14 (2012-03-13)
Polychlorinated biphenyls (PCBs) are still prevalent in the environment despite the fact that they have been banned in many countries for several decades. Recent epidemiologic studies have demonstrated a link between PCBs exposure and pathological alterations of bone tissues. The
L A Martin et al.
Drug metabolism and disposition: the biological fate of chemicals, 40(3), 588-595 (2011-12-22)
Polychlorinated biphenyl (PCB) congeners differentially reduce serum thyroxine (T(4)) in rats, but little is known about their ability to affect biliary excretion of T(4). Thus, male Sprague-Dawley rats were orally administered Aroclor-1254, Aroclor-1242 (32 mg/kg per day), PCB-95, PCB-99, PCB-118
Jiali Cai et al.
Toxicology, 287(1-3), 21-28 (2011-06-15)
The testicular toxicity of polychlorinated biphenyls (PCBs) has been extensively studied. However, the detailed mechanism is still obscure. In the present study, male C57 mice were treated with different doses of Aroclor 1254 (a commercial PCB mixture) once every 3
Lidia Strużyńska et al.
Toxicology, 300(1-2), 12-18 (2012-05-26)
Aroclor 1254, a commercially produced mixture of polychlorinated biphenyls, is known to cause many adverse conditions, including neurotoxicity. It has been recently postulated that upregulation of N-methyl-d-aspartate receptors (NMDARs) and enhanced glutamate signalling which leads to excitotoxicity, is the mechanism

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service