Skip to Content
Merck
All Photos(1)

Key Documents

47943

Supelco

ASTM® D5441 MTBE Contaminants (low) Mix B

0.1 % (w/w) each component in methyl tert-butyl ether, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€51.10

€51.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€51.10

About This Item

UNSPSC Code:
12000000

€51.10


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Agency

ASTM® D5441

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

0.1 % (w/w) each component in methyl tert-butyl ether

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-8°C

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    tert-Amyl methyl ether
    tert-Butanol
    tert-Butyl ethyl ether
    Methanol
    2-Methylbutane
    2-Methyl-2-butene
    Pentane
    cis-2-Pentene
    trans-2-Pentene
    Triisobutylene
    2,4,4-Trimethyl-1-pentene
    See All (11)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-27.4 °F - closed cup

Flash Point(C)

-33.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service