Skip to Content
Merck
All Photos(1)

Key Documents

47884

Supelco

Isomaltotriose, DP3

analytical standard

Synonym(s):

Isomaltotriose, α-D-Glucosyl-[1→6]-α-D-glucosyl-[1→6]-D-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O16
CAS Number:
Molecular Weight:
504.44
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

feature

standard type oligosaccharide

analyte chemical class(es)

oligosaccharides

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-30°C

SMILES string

OC[C@H]1O[C@H](OC[C@H]2O[C@H](OC[C@H]3OC(O)[C@H](O)[C@@H](O)[C@@H]3O)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C18H32O16/c19-1-4-7(20)11(24)14(27)17(33-4)31-3-6-9(22)12(25)15(28)18(34-6)30-2-5-8(21)10(23)13(26)16(29)32-5/h4-29H,1-3H2/t4-,5-,6-,7-,8-,9-,10+,11+,12+,13-,14-,15-,16?,17+,18+/m1/s1

InChI key

FBJQEBRMDXPWNX-FYHZSNTMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of methyl alpha-isomalto-oligosaccharides specifically deoxygenated at position 2 of the terminal glycopyranosyl unit.
E Petráková et al.
Glycoconjugate journal, 11(1), 17-22 (1994-02-01)
J J Bourgarit et al.
Biochimie, 66(11-12), 711-715 (1984-11-01)
A methodology for the pharmacodynamical assay of mixtures of 2 spasmogenic peptides is described, illustrated and discussed, with reference to a method intended for the chemists (P. Job, 1927). This allows a quick appreciation of the existence of synergism, of
E Shimizu et al.
Bioscience, biotechnology, and biochemistry, 62(1), 117-122 (1998-03-21)
An isomaltotriose-producing endo-dextranase was simply purified from cell-free culture broth of a Fusarium sp. by ethanol fractionation and consecutive column chromatographies using DEAE-Toyopearl and Bio-Gel P-100. The purified enzyme was judged to be homogeneous on PAGE and SDS-PAGE as well
R E McLaughlin et al.
FEMS microbiology letters, 140(2-3), 261-264 (1996-07-01)
The multiple-sugar metabolism (msm) locus of Streptococcus mutans constitutes a non-PTS sugar uptake system responsible for the transport and utilization of raffinose, melibiose and isomaltotrioses. While previous studies have used polar mutations to suggest that these genes are co-transcribed, there
R R Russell et al.
The Journal of biological chemistry, 267(7), 4631-4637 (1992-03-05)
An 11-kilobase gene region of Streptococcus mutans has been identified which contains eight contiguous genes involved with the uptake and metabolism of multiple sugars (the msm system). Sequence analysis of this region indicates that several of these genes specify proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service