47743-U
ASTM® D5580 Quantitative Cal Mix 4
(Each component at nominal wt.% indicated.), analytical standard
About This Item
Recommended Products
grade
analytical standard
Agency
ASTM® D5580
packaging
ampule of 1 mL
concentration
(Each component at nominal wt.% indicated.)
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
petroleum
format
multi-component solution
storage temp.
room temp
Application
Legal Information
Analyte
Signal Word
Danger
Hazard Statements
Precautionary Statements
Flash Point(F)
10.0 °F
Flash Point(C)
-12.2 °C
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service