Skip to Content
Merck
All Photos(1)

Key Documents

47616-U

Supelco

Toxaphene solution

analytical standard, 200 μg/mL in isooctane, ampule of 1 mL

Synonym(s):

Camphechlor solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
77101502
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

feature

calibration

packaging

ampule of 1 mL

concentration

200 μg/mL in isooctane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

room temp

InChI

1S/C10H8Cl8/c1-4-7(2-11,3-12)9(16)6(14)5(13)8(4,15)10(9,17)18/h5-6H,1-3H2

InChI key

OEJNXTAZZBRGDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F

Flash Point(C)

-12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toxaphene (polychlorinated camphenes).
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 20, 327-348 (1979-10-01)
M A Saleh
Reviews of environmental contamination and toxicology, 118, 1-85 (1991-01-01)
The chemistry of toxaphene is now well developed; 20 isomers have been isolated and identified. The molecular weight and molecular formula are known for the remaining major components. The major metabolic degradation mechanisms for toxaphene in all organisms from bacteria
John R Kucklick et al.
Analytical and bioanalytical chemistry, 386(4), 819-836 (2006-10-05)
Recent advances in the analysis of the chlorinated environmental pollutants polychlorinated naphthalenes (PCNs) and toxaphene are highlighted in this review. Method improvements have been realized for PCNs over the past decade in isomer-specific quantification, peak resolution, and the availability of
H J de Geus et al.
Environmental health perspectives, 107 Suppl 1, 115-144 (1999-05-07)
Toxaphene production, in quantities similar to those of polychlorinated biphenyls, has resulted in high toxaphene levels in fish from the Great Lakes and in Arctic marine mammals (up to 10 and 16 microg g-1 lipid). Because of the large variabiliity
Toxaphene.
G A Pollock et al.
Residue reviews, 69, 87-140 (1978-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service