Skip to Content
Merck
All Photos(1)

Key Documents

47120-U

Supelco

Safflower oil

analytical standard

Synonym(s):

Safflower seed oil from Carthamus tinctorius seed

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
85151701

biological source

plant seeds (Carthamus tinctorius)

grade

analytical standard

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.476 (lit.)

density

0.921 g/mL at 25 °C (lit.)

application(s)

food and beverages

format

neat

functional group

carboxylic acid
oleic acid

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

This reference material is offered as a qualitative standard. Use the chromatographic fingerprint of our reference material to check for the presence of safflower oil in your sample. A Certificate of Composition which includes a chromatogram is shipped with each product.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

12 - Non Combustible Liquids

WGK

awg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Ji et al.
American journal of physiology. Gastrointestinal and liver physiology, 302(11), G1292-G1300 (2012-03-31)
Previous studies have linked certain types of gut mucosal immune cells with fat intake. We determined whether fat absorption activates intestinal mucosal mast cells (MMC), a key component of the gut mucosal immune system. Conscious intestinal lymph fistula rats were
Melisa Kurtz et al.
Molecular and cellular endocrinology, 362(1-2), 120-127 (2012-06-13)
The fetal lung is affected by maternal diabetes. Nuclear receptor PPARα regulates nitric oxide (NO) overproduction in different tissues. We aimed to determine whether fetal lung PPARα expression is altered by maternal diabetes, and if there are gender-dependent changes in
Letter to the editor.
Lawrence L Rudel
Journal of clinical lipidology, 5(6), 500-500 (2011-11-24)
Eric E Noreen et al.
Journal of complementary & integrative medicine, 9, doi:10-doi:10 (2012-10-30)
To determine the effects of 6wk of supplementation with fish oil (FO) on blood pressure and the morning salivary cortisol concentration in normotensive adults. Testing was performed following an overnight fast. Subjects (n=40; 35+/-13y, mean+/-SD) rested supine for 40min, at
K G Lamping et al.
Diabetes, obesity & metabolism, 15(2), 144-152 (2012-09-07)
The ability of dietary enrichment with monounsaturated fatty acid (MUFA), n-3 or n-6 polyunsaturated fatty acids (PUFAs) to reverse glucose intolerance and vascular dysfunction resulting from excessive dietary saturated fatty acids is not resolved. We hypothesized that partial replacement of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service