Skip to Content
Merck
All Photos(1)

Key Documents

442876

Supelco

Bisphenol A-d16

analytical standard

Synonym(s):

2,2-Bis(4-hydroxyphenyl)propane-d16

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2C(C6D4OD)2
CAS Number:
Molecular Weight:
244.38
UNSPSC Code:
23151816
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 50 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

220 °C/4 mmHg (lit.)

mp

157-159 °C (lit.)

application(s)

environmental

format

neat

mass shift

M+16

storage temp.

2-30°C

SMILES string

[2H]Oc1c([2H])c([2H])c(c([2H])c1[2H])C(c2c([2H])c([2H])c(O[2H])c([2H])c2[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17H,1-2H3/i1D3,2D3,3D,4D,5D,6D,7D,8D,9D,10D/hD2

InChI key

IISBACLAFKSPIT-MAJJRYNQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Repr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Alkylphenols are used in various industries like paper manufacturing and pesticide formulations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service