Skip to Content
Merck
All Photos(1)

Documents

442867

Supelco

Dipentyl phthalate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O4
CAS Number:
Molecular Weight:
306.40
Beilstein:
1987323
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

1.025 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

CCCCCOC(=O)c1ccccc1C(=O)OCCCCC

InChI

1S/C18H26O4/c1-3-5-9-13-21-17(19)15-11-7-8-12-16(15)18(20)22-14-10-6-4-2/h7-8,11-12H,3-6,9-10,13-14H2,1-2H3

InChI key

IPKKHRVROFYTEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dipentyl phthalate belongs to the group of phthalates, widely used as plasticizers in plastics and for consumer applications. It is a major contaminant and can be released from a variety of products into the environment.

Application

Dipentyl phthalate may be used as an analytical reference standard for the quantification of the analyte in water, juice and milk samples using high-performance liquid chromatography (HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Graphene reinforced hollow fiber liquid-phase microextraction for the determination of phthalates in water, juice and milk samples by HPLC
Sun M, et al.
Analytical Methods : Advancing Methods and Applications, 5(20), 5694-5700 (2013)
Ji-Young Ahn et al.
Current microbiology, 52(5), 340-344 (2006-04-06)
The efficiency of two lipolytic enzymes (fungal cutinase and yeast esterase) in the degradation of dipentyl phthalate (DPeP) was investigated. The DPeP degradation rate of fungal cutinase was surprisingly high, i.e., almost 60% of the initial DPeP (500 mg/L) was
N P Moore et al.
Archives of toxicology, 66(6), 435-442 (1992-01-01)
Cell-specific testicular toxicants have been used to examine the distribution of creatine within the rat testis, and to examine the potential use of creatinuria as a non-invasive indicator of testicular damage. Groups of rats were administered single doses of various
J J Heindel et al.
Toxicology and applied pharmacology, 97(2), 377-385 (1989-02-01)
High doses of phthalate esters in vivo cause testicular lesions. One initial target for these effects is the sustentacular Sertoli cell. Sertoli cells are unique in that they have surface membrane receptors for follicle-stimulating hormone (FSH), which couple to adenylate
D M Creasy et al.
Experimental and molecular pathology, 46(3), 357-371 (1987-06-01)
A sequential morphological study has been carried out to examine the ultrastructural effects of di-n-pentyl phthalate (DPP) on the mature rat testis. A single oral dose of 2.2 g DPP/kg body wt was administered, and testes, perfuse-fixed 3-48 hr after

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service