Skip to Content
Merck
All Photos(1)

Documents

442578

Supelco

Endrin aldehyde

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12000000

grade

analytical standard

product line

PESTANAL®

CofA

current certificate can be downloaded

packaging

ampule of 25 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

neat

storage temp.

2-30°C

InChI

1S/C12H8Cl6O/c13-8-5-3(2-19)1-4-6(5)9(14,12(8,17)18)11(16)7(4)10(8,11)15/h2-7H,1H2

InChI key

HCTWZIFNBBCVGM-UHFFFAOYSA-N

General description

Endrin aldehyde can be obtained as an oxidation product of the pesticide endrin, upon exposure to light or heat.
Endrin aldehyde is an organochlorine type of pesticide.

Application

Endrin aldehyde may be used as a reference standard for the analysis of endrin aldehyde in:
  • Airborne particulate matter samples by using ultrasound assisted extraction micro-scale cell (UAE-MSC) and gas chromatography/mass spectrometry in negative chemical ionization (GC/MS-NCI) mode.
  • Seawater samples by solid-phase extraction (SPE) and UAE followed by analysis using GC coupled to triple quadrupole tandem mass spectrometry (MS/MS).
  • Brackish water fish by GC-MS.
  • Biological matrices by quick, easy, cheap, effective, rugged, and safe (QuEChERS) extraction and GC-MS/MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Levels of Organochlorine Pesticides in Brackish Water Fish from Niger River, Nigeria.
Unyimadu JP, et al.
Journal of Environmental and Public Health, 2018 (2018)
Gas chromatography-vacuum ultraviolet spectroscopy for multiclass pesticide identification.
Fan H, et al.
Journal of Chromatography A, 1389, 120-127 (2015)
Patnaik P
Handbook of Environmental Analysis: Chemical Pollutants in Air, Water, Soil, and Solid Wastes, Third Edition (2017)
Patnaik P
A Comprehensive Guide to the Hazardous Properties of Chemical Substances (2007)
Multi-residue analysis of legacy POPs and emerging organic contaminants in Singapore's coastal waters using gas chromatography-triple quadrupole tandem mass spectrometry.
Zhang H, et al.
The Science of the Total Environment, 523, 219-232 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service