Skip to Content
Merck
All Photos(1)

Key Documents

442523

Supelco

Chrysene-d12

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18D12
CAS Number:
Molecular Weight:
240.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 25 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

448 °C (lit.)

mp

252-254 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

mass shift

M+12

storage temp.

2-30°C

SMILES string

[2H]c1c([2H])c([2H])c2c(c1[2H])c([2H])c([2H])c3c4c([2H])c([2H])c([2H])c([2H])c4c([2H])c([2H])c23

InChI

1S/C18H12/c1-3-7-15-13(5-1)9-11-18-16-8-4-2-6-14(16)10-12-17(15)18/h1-12H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D

InChI key

WDECIBYCCFPHNR-AQZSQYOVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xiaowei Wang et al.
Journal of separation science, 37(14), 1842-1849 (2014-04-29)
An automatic exposed and in-syringe dynamic single-drop microextraction method (SDME) for the determination of five polycyclic musks in natural waters was developed using gas chromatography with mass spectrometry. Online agitation was first introduced to the automatic SDME with a magnetic
Zoe Ruge et al.
Environmental science & technology, 49(23), 13777-13786 (2015-10-06)
Polycyclic aromatic hydrocarbons (PAHs) and polybrominated diphenylethers (PBDEs) are both currently released into the environment from anthropogenic activity. Both are hence primarily associated with populated or industrial areas, although wildfires can be an important source of PAHs, as well. Polyethylene
Jian-Lin Sun et al.
Ecotoxicology and environmental safety, 113, 31-37 (2014-12-09)
Halogenated polycyclic aromatic hydrocarbons (HPAHs) have been reported to occur widely in urban air. Nevertheless, knowledge about the human health risk associated with inhalation exposure to HPAHs is scarce so far. In the present study, nine HPAHs and 16 PAHs
Kenshi Sankoda et al.
Archives of environmental contamination and toxicology, 68(2), 217-224 (2014-12-07)
This study aimed to investigate the behavior of organic ultraviolet (UV) filters released by recreational activities along the Japanese coastline. Seasonal variations of organic UV filters in seawater were investigated at four different recreational beaches (Mogushi, Wakamiya, Tsurugahama, and Otachimisaki
Badreddine Barhoumi et al.
Marine environmental research, 101, 184-195 (2014-08-12)
In this study, biological responses and contaminant levels in biological tissues were investigated in grass goby fish specimens (Zosterisessor ophiocephalus) collected from five stations in a moderately polluted ecosystem, namely the Bizerte lagoon on the north coast of Tunisia. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service