Skip to Content
Merck
All Photos(1)

Documents

442441

Supelco

Acrolein-2,4-DNPH

analytical standard

Synonym(s):

Acrolein-2,4-dinitrophenylhydrazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N4O4
CAS Number:
Molecular Weight:
236.18
UNSPSC Code:
12000000

grade

analytical standard

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 25 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-30°C

InChI

1S/C9H8N4O4/c1-2-5-10-11-8-4-3-7(12(14)15)6-9(8)13(16)17/h2-6,11H,1H2/b10-5+

InChI key

JJPZHGIYUVFTGG-BJMVGYQFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acrolein-2,4-DNPH is a derivative of acrolein formed via chemical reaction of acrolein with 2,4-dinitrophenylhydrazine in the presence of trifluoroacetic acid.

Application

Acrolein-2,4-DNPH has been used to determine the amount of acrolein present in polyethylene combustion product (PCP) extract, using high performance liquid chromatography (HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toxicity of smoke extracts towards A549 lung cells: role of acrolein and suppression by carbonyl scavengers
Burcham.CP, et al.
Chemico-Biological Interactions, 183, 416-424 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service