Skip to Content
Merck
All Photos(1)

Key Documents

442418

Supelco

4-Methylphenol

analytical standard

Synonym(s):

p-Cresol, 4-Methylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
1305151
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

3.72 (vs air)

vapor pressure

1 mmHg ( 20 °C)

autoignition temp.

1038 °F

expl. lim.

1 %
1.1 %, 150 °F

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

202 °C (lit.)

mp

32-34 °C (lit.)

density

1.034 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

storage temp.

room temp

SMILES string

Cc1ccc(O)cc1

InChI

1S/C7H8O/c1-6-2-4-7(8)5-3-6/h2-5,8H,1H3

InChI key

IWDCLRJOBJJRNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methylphenol is an isomer of the phenolic compound, named cresol. It can be formed via action of colonic bacteria on tyrosine and phenylalanine in human body. It can act like a an odorant and a component of human sweat, which can have the tendency to attract female Anopheles mosquitoes, the world′s most important vector of Plasmodium falciparum malaria.

Application

4-Methylphenol can be used in the preparation of toluene via hydrodeoxygenation in the presence of cobalt dispersed on molybdenum disulfide monolayers as catalyst.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olfaction: mosquito receptor for human-sweat odorant
Hallema.AE, et al.
Nature, 427, 212-213 (2004)
MoS2 monolayer catalyst doped with isolated Co atoms for the hydrodeoxygenation reaction
Liu G, et al.
Nature Chemistry (2017)
Chronic Kidney Disease, Dialysis, and Transplantation E-Book: A Companion to Brenner and Rector?s The Kidney (2010)
Hydrogenation of o-cresol on platinum catalyst: Catalytic experiments and first-principles calculations
Li Y, et al.
Applied Surface Science, 393, 212-220 (2017)
K Akasaka et al.
Journal of chromatography, 617(2), 205-211 (1993-08-11)
Cholesterol ester (ChE) and triacylglycerol (TG) hydroperoxides in human plasma were determined by high-performance liquid chromatography with postcolumn detection with diphenyl-1-pyrenylphosphine. Human plasma was extracted once with n-hexane. 2,6-Di-tert.-butyl-4-methylphenol and N-stearylcinnamide were added to human plasma before extraction as an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service