Skip to Content
Merck
All Photos(1)

Key Documents

442391

Supelco

3-Methylphenol

analytical standard

Synonym(s):

m-Cresol, 1-Hydroxy-3-methylbenzene, 3-Hydroxytoluene, 3-Methylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
506719
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

vapor density

3.72 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

CofA

current certificate can be downloaded

autoignition temp.

1036 °F

expl. lim.

1.06-1.35 %, 150 °F

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.541 (lit.)

bp

203 °C (lit.)

mp

8-10 °C (lit.)

density

1.034 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

storage temp.

2-30°C

SMILES string

Cc1cccc(O)c1

InChI

1S/C7H8O/c1-6-3-2-4-7(8)5-6/h2-5,8H,1H3

InChI key

RLSSMJSEOOYNOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methylphenol, a substituted phenol, is a common water pollutant. It is found to be toxic to fish and aquatic biota, even when present in trace quantities.[1][2]

Application

2-Methylphenol may be used as an analytical reference standard for the determination of the analyte in petroleum crude oils,[3][4] industrial wastewater[1] and in fish bile[5] by various chromatographic techniques.
3-Methylphenol can be used in the preparation of 3-methylcyclohexanone via hydrogenation reaction in the presence of nickel catalyst.[6] It can also be used in the synthesis of toluene, via hydrogenation in the presence of bimetallic nickel-iron catalyst.[6] It can find applications as a solvent for the ultraviolet-visible light spectral studies of chemically synthesized soluble acrylic acid doped polyaniline.[7]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Selective conversion of m-cresol to toluene over bimetallic Ni?Fe catalysts
Nie L, et al.
J. Mol. Catal. A: Chem., 388, 47-55 (2014)
Studies on chemically synthesized soluble acrylic acid doped polyaniline
Athawale.AA, et al.
Materials Chemistry and Physics, 73, 106-110 (2002)
Determination of phenols in petroleum crude oils using liquid chromatography with electrochemical detection
MacCrehan WA and Brown-Thomas JM
Analytical Chemistry, 59(3), 477-479 (1987)
Optimization of isocratic mobile phase composition for HPLC analysis of eleven substituted phenols
Busto O, et al.
Chromatographia, 32(11-12), 566-572 (1991)
Identification and analysis of C0-C3 phenols in some Australian crude oils
Ioppolo M, et al.
Organic Geochemistry, 18(5), 603-609 (1992)

Protocols

Analytical standard separation of various phenols for research and industrial applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service