Skip to Content
Merck
All Photos(1)

Key Documents

442377

Supelco

3,5-Di-tert-butyl-4-hydroxytoluene

analytical standard

Synonym(s):

2,6-Di-tert-butyl-4-methylphenol, 2,6-Di-tert-butyl-p-cresol, BHT, Butylated hydroxytoluene, Butylhydroxytoluene, DBPC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H2(CH3)OH
CAS Number:
Molecular Weight:
220.35
Beilstein:
1911640
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

7.6 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

autoignition temp.

878 °F

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

265 °C (lit.)

mp

69-73 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

SMILES string

Cc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3

InChI key

NLZUEZXRPGMBCV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

260.6 °F - open cup

Flash Point(C)

127 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Vijesh et al.
European journal of medicinal chemistry, 46(11), 5591-5597 (2011-10-05)
In the present study two new series of Hantzsch 1,4-dihydropyridine derivatives (1,4-DHPs) containing substituted pyrazole moiety (4a-f and 5a-f) were synthesized by the reaction of 3-aryl-1H-pyrazole-4-carbaldehydes with 1,3-dicarbonylcompounds (ethylacetoacetate and methylacetoacetate) and ammonium acetate. The newly synthesized compounds were characterized
Donatella Boschi et al.
Journal of medicinal chemistry, 49(10), 2886-2897 (2006-05-12)
The synthesis and study of the antioxidant and vasodilator properties of a new class of phenols able to release nitric oxide are described. The products were designed through a symbiotic approach using selected phenols and selected nitrooxy and furoxan NO-donors
Xiao-Juan Duan et al.
Journal of natural products, 70(7), 1210-1213 (2007-07-03)
Four new highly brominated and fully substituted mono- and bis-phenols, 1-(2,3,6-tribromo-4,5-dihydroxybenzyl)pyrrolidin-2-one (1), 1,2-bis(2,3,6-tribromo-4,5-dihydroxyphenyl)ethane (2), 6-(2,3,6-tribromo-4,5-dihydroxybenzyl)-2,5-dibromo-3,4-dihydroxybenzyl methyl ether (3), and 2,3,6-tribromo-4,5-dihydroxybenzyl methyl sulfone (4), were characterized from the marine red alga Symphyocladia latiuscula. In addition, five known bromophenols, bis(2,3,6-tribromo-4,5-dihydroxyphenyl)methane (5), bis(2,3,6-tribromo-4,5-dihydroxybenzyl)
Canan Kuş et al.
Bioorganic & medicinal chemistry, 16(8), 4294-4303 (2008-03-14)
Some novel 1-methyl-4-(2-(2-substitutedphenyl-1H-benzimidazol-1-yl)acetyl)thiosemicarbazides (16a-20a), 5-[(2-(substitutedphenyl)-1H-benzimidazol-1-yl)methyl]-N-methyl-1,3,4-thiadiazol-2-amines (17b-20b), and 5-[(2-(substitutedphenyl)-1H-benzimidazol-1-yl)methyl-4-methyl-2H-1,2,4-triazole-3(4H)-thiones (16c-20c) were synthesized and tested for antioxidant properties by using various in vitro systems. Compounds 16a-20a were found to be a good scavenger of DPPH radical (IC(50), 26 microM; IC(50), 30 microM;
Murat Sentürk et al.
Bioorganic & medicinal chemistry, 17(8), 3207-3211 (2009-02-24)
The inhibition of two human cytosolic carbonic anhydrase (hCA, EC 4.2.1.1) isozymes I and II, with a series of phenol derivatives was investigated by using the esterase assay, with 4-nitrophenyl acetate as substrate. 2,6-Dimethylphenol, 2,6-diisopropylphenol (propofol), 2,6-di-t-butylphenol, butylated hydroxytoluene, butylated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service