Skip to Content
Merck
All Photos(1)

Key Documents

442210

Supelco

1,1-Dichloro-2-propanone

analytical standard

Synonym(s):

1,1-Dichloroacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCHCl2
CAS Number:
Molecular Weight:
126.97
Beilstein:
1740214
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.446 (lit.)

bp

117-118 °C (lit.)

density

1.327 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

room temp

SMILES string

CC(=O)C(Cl)Cl

InChI

1S/C3H4Cl2O/c1-2(6)3(4)5/h3H,1H3

InChI key

CSVFWMMPUJDVKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.4 °F - closed cup

Flash Point(C)

45.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F B Daniel et al.
Drug and chemical toxicology, 16(3), 293-305 (1993-01-01)
Groups of 10 male and 10 female Sprague-Dawley rats were administered 1,1-dichloro-2-propanone in corn oil by gavage at 0, 10, 20, 40, or 80 mg/kg/day for 90 consecutive days. Food and water consumption, body and organ weights, organ-to-body weight ratios
Xu Xu et al.
Journal of exposure analysis and environmental epidemiology, 15(4), 289-296 (2004-08-19)
Dermal contact with some organic disinfection by-products (DBPs) such as trihalomethanes in chlorinated drinking water has been established to be an important exposure route. We evaluated dermal absorption of two haloketones (1,1-dichloropropanone and 1,1,1-trichloropropanone) and chloroform while bathing, by collecting
Nan Huang et al.
Water research, 114, 246-253 (2017-03-04)
Benzalkonium chlorides (BACs), as typical cationic surfactants and biocides widely applied in household and industrial products, have been frequently detected as micropollutants in many aquatic environments. In this study, the combination of UV irradiation and chlorine (UV/chlorine), a newly interested
Yishuai Pan et al.
Chemosphere, 224, 228-236 (2019-03-02)
Metronidazole (MET) is a widely used antibiotic but is recalcitrant in aquatic environment. This study investigated elimination of MET by UV/chlorine process systematically. The degradation of MET in the process well fitted pseudo first-order kinetics. Decreasing pH from 9 to
Yu Li et al.
Chemosphere, 252, 126611-126611 (2020-05-24)
Halogenated disinfection byproducts (DBPs) are formed during chlorine disinfection of drinking water. The complicated natural organic matter in source water causes the formation of an even more complicated mixture of DBPs. To evaluate the toxicity of a DBP mixture in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service