Skip to Content
Merck
All Photos(1)

Key Documents

41089

Supelco

Alachlor solution

certified reference material, 1000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20ClNO2
CAS Number:
Molecular Weight:
269.77
Beilstein:
2944476
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

CCc1cccc(CC)c1N(COC)C(=O)CCl

InChI

1S/C14H20ClNO2/c1-4-11-7-6-8-12(5-2)14(11)16(10-18-3)13(17)9-15/h6-8H,4-5,9-10H2,1-3H3

InChI key

XCSGPAVHZFQHGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Alachlor is an acetanilide[1], pre-emergence herbicide, which can find a broad spectrum of applications in agriculture[2], for weed control management[3]. It can undergo photo-degradation via photo-Fenton process, a combination of Fenton reagent with ultraviolet radiations.[3]

Application

Alachlor is an acetanilide[4], pre-emergence herbicide, which can find a broad spectrum of applications in agriculture[5], for weed control management[6]. It can undergo photo-degradation via photo-Fenton process, a combination of Fenton reagent with ultraviolet radiations.[6]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - Methanol

Flash Point(C)

9.7 °C - Methanol


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improving alachlor biodegradability by ferrate oxidation
Zhu J-H, et al.
Journal of Hazardous Materials, 135, 94-99 (2006)
Improving alachlor biodegradability by ferrate oxidation
Zhu J-H, et al.
Journal of Hazardous Materials, 135(1), 94-99 (2006)
Photo-Fenton degradation of alachlor in the presence of citrate solution
Katsumata H, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 180(1), 38-45 (2006)
Photo-Fenton degradation of alachlor in the presence of citrate solution
Katsumata H, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 180, 38-45 (2006)
Degradation mechanism of alachlor during direct ozonation and O 3/H 2 O 2 advanced oxidation process
Qiang Z, et al.
Chemosphere, 78(5), 517-526 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service