Skip to Content
Merck
All Photos(1)

Key Documents

40862

Supelco

Flavone

analytical standard

Synonym(s):

2-Phenyl-4H-1-benzopyran-4-one, 2-Phenylchromone

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€51.90

€51.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€51.90

About This Item

Empirical Formula (Hill Notation):
C15H10O2
CAS Number:
Molecular Weight:
222.24
Beilstein:
157598
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€51.90


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

94-97 °C (lit.)

SMILES string

O=C1C=C(Oc2ccccc12)c3ccccc3

InChI

1S/C15H10O2/c16-13-10-15(11-6-2-1-3-7-11)17-14-9-5-4-8-12(13)14/h1-10H

InChI key

VHBFFQKBGNRLFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service