Skip to Content
Merck
All Photos(1)

Key Documents

40220-U

Supelco

Aldrin solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8Cl6
CAS Number:
Molecular Weight:
364.91
Beilstein:
2336652
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

ClC1=C(Cl)[C@]2(Cl)[C@@H]3[C@@H]4C[C@@H](C=C4)[C@@H]3[C@@]1(Cl)C2(Cl)Cl

InChI

1S/C12H8Cl6/c13-8-9(14)11(16)7-5-2-1-4(3-5)6(7)10(8,15)12(11,17)18/h1-2,4-7H,3H2/t4-,5+,6+,7-,10+,11-

InChI key

QBYJBZPUGVGKQQ-SJJAEHHWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup - Solvent

Flash Point(C)

9.7 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hans Ragnar Norli et al.
Journal of chromatography. A, 1218(41), 7234-7241 (2011-09-09)
The QuEChERS method developed for 22 organochlorine pesticides (OCPs) and 7-polychlorinated biphenyls (PCBs) in fish tissue involves a simple and efficient freezing technique for removal of lipids. The equipment developed consists of disposable syringes and a freezing block constructed from
Daibin Yang et al.
Environmental science & technology, 44(5), 1854-1859 (2010-02-11)
alpha-HCH (hexachlorocyclohexane) is chiral and can still be detected in almost all environmental media. In this study, the enantioselective behavior of alpha-HCH in mice (CD1) and quail (Coturnix japonica) was investigated and compared after a single dose of exposure. The
Glenn A Rowland et al.
Environmental pollution (Barking, Essex : 1987), 159(5), 1076-1084 (2011-03-15)
Organochlorine (OC) contaminants are transported to the Polar Regions, where they have the potential to bioaccumulate, presenting a threat to the health of wildlife and indigenous communities. They deposit onto snowpack during winter, and accumulate until spring, when they experience
A Dvorská et al.
Environmental science and pollution research international, 19(6), 1944-1952 (2012-07-07)
Organochlorinated pesticides were widely applied in Armenia until the 1980s, like in all former Soviet Union republics. Subsequently, the problem of areas contaminated by organochlorinated pesticides emerged. Environmental, waste and food samples at one pesticide burial site (Nubarashen) and three
Xiao-Yuan Tao et al.
Molecular ecology, 21(17), 4371-4385 (2012-04-21)
Cotton plants accumulate phytotoxins, including gossypol and related sesquiterpene aldehydes, to resist insect herbivores and pathogens. To counteract these defensive plant secondary metabolites, cotton bollworms (Helicoverpa armigera) elevate their production of detoxification enzymes, including cytochrome P450 monooxygenases (P450s). Besides their

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service