Skip to Content
Merck
All Photos(1)

Key Documents

40051

Supelco

Hexachlorocyclopentadiene solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

Agency

EPA 508

product line

TraceCERT®

feature

standard type calibration

packaging

ampule of 1 mg

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C5Cl6/c6-1-2(7)4(9)5(10,11)3(1)8

InChI key

VUNCWTMEJYMOOR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexachlorocyclopentadiene is an organochlorine compound, which may be used, in the manufacture of some pesticides.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sublethal effects of the flame retardant intermediate hexachlorocyclopentadiene (HCCPD) on the gene transcription and protein activity of Daphnia magna
Houde M, et al.
Aquatic Toxicology (Amsterdam, Netherlands), 140, 213-219 (2013)
Teratogenic potential of hexachlorocyclopentadiene in mice and rabbits.
F J Murray et al.
Toxicology and applied pharmacology, 53(3), 497-500 (1980-05-01)
Fate of hexachlorocyclopentadiene in water and goldfish.
A A Podowski et al.
Archives of environmental contamination and toxicology, 13(4), 471-481 (1984-07-01)
S M El Dareer et al.
Journal of toxicology and environmental health, 12(2-3), 203-211 (1983-08-01)
The differential disposition of hexachlorocyclopentadiene (HCCP) following oral administration, as contrasted to inhalation or intravenous administration, may account for its lower toxicity by this route. Following an intravenous dose of [14C]HCCP to rats at 0.59 mg/kg, 39.0% of the radioactivity
Hexachlorocyclopentadiene contamination of a municipal wastewater treatment plant.
J R Kominsky et al.
American Industrial Hygiene Association journal, 41(8), 552-556 (1980-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service