Skip to Content
Merck
All Photos(1)

Key Documents

40013

Supelco

1,1,2-Trichloroethane solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

InChI

1S/C2H3Cl3/c3-1-2(4)5/h2H,1H2

InChI key

UBOXGVDOUJQMTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ariel Grostern et al.
Applied and environmental microbiology, 72(1), 428-436 (2006-01-05)
Mixed anaerobic microbial subcultures enriched from a multilayered aquifer at a former chlorinated solvent disposal facility in West Louisiana were examined to determine the organism(s) involved in the dechlorination of the toxic compounds 1,2-dichloroethane (1,2-DCA) and 1,1,2-trichloroethane (1,1,2-TCA) to ethene.
Robert Zaleśny et al.
Journal of molecular modeling, 13(6-7), 785-791 (2007-04-18)
The UV-Vis spectra of series of polymethylmethacrylate (PMMA) copolymers with attached trans-azobenzene derivatives were measured in 1,1,2-trichloroethane. In order to gain some insight into the recorded spectra, the quantum chemical calculations were performed for the substituted azobenzenes using both configuration
Michio M Matsushita et al.
Journal of the American Chemical Society, 127(36), 12450-12451 (2005-09-08)
A bromide salt of a TTF-based cross-cyclophane-type donor displayed characteristic nonlinear voltage-biased conductivity. As the applied voltage was increased, the passing current was enhanced abruptly at 54 kV.cm-1, exhibiting hysteretic behavior. X-ray analysis of the current-induced low-resistance state of the
1,1,2-trichloroethane.
IARC monographs on the evaluation of carcinogenic risks to humans, 52, 337-359 (1991-01-01)
Chuanjie Cheng et al.
The Journal of organic chemistry, 74(15), 5671-5674 (2009-06-19)
In the presence of 1,1,2-trichloroethane, a novel procedure for the Pd-C catalytic N-debenzylation of benzylamines was established. The method proceeded in a synergistic catalytic system and directly gave the products as crystal amine hydrochlorides in practically quantitative yields.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service