Skip to Content
Merck
All Photos(1)

Key Documents

33143

Supelco

Vials, crimp top, serum bottle

volume 10 mL, amber glass bottle, O.D. × H 25 mm × 50 mm, pkg of 36 ea

Sign Into View Organizational & Contract Pricing

Select a Size

36 EA
€75.40

€75.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
36 EA
€75.40

About This Item

UNSPSC Code:
41121806

€75.40


Please contact Customer Service for Availability

Request a Bulk Order

material

amber glass bottle

feature

autoclavable
closure type crimp top vial

packaging

pkg of 36 ea

O.D. × H

25 mm × 50 mm

volume

10 mL

fitting

thread for 20 mm

Looking for similar products? Visit Product Comparison Guide

General description

These vials are constructed of clear, borosilicate glass; closure not included.

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service