Skip to Content
Merck
All Photos(1)

Key Documents

28861-U

Supelco

Super Clean (In-Line Design) Gas Purifier

moisture trap, stainless steel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
SB.54

material

stainless steel

Looking for similar products? Visit Product Comparison Guide

General description

Installation and replacement of the Click-On purifier is simple and eliminates the risk of damage to the gas line connections caused by overtightening. Simply attach the connectors to the gas line, install the purifier, and hand tighten the connectors.

Note: Does not include Click-on Connectors, these must be purchased before initial use.

Dimensions:
  • Cartridge and Connectors: 11 1/2 in. long x 1 1/4 in. diameter
  • Cartridge Only: 7 5/8 in. long x 1 1/4 in. diameter

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service