Key Documents
27439
Supelco
Screw cap
black polypropylene (Top Hat), PTFE/silicone, for use with 4 mL vial with 13-425 thread, pkg of 100 ea
Select a Size
100 EA
€75.10
€75.10
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
100 EA
€75.10
About This Item
UNSPSC Code:
41122001
€75.10
Please contact Customer Service for Availability
Request a Bulk Order
material
PTFE/silicone
black polypropylene (Top Hat)
feature
pre-assembled
packaging
pkg of 100 ea
fitting
top hat cap
compatibility
for use with 4 mL vial with 13-425 thread
General description
Preassembled closure and septa designed to provide a positive physical lock to assure that components arrive intact.
Application
PTFE-silicone-PTFE screw cap may be used for capping samples contained in a micro-insert vial, to be loaded on to GC/MS for biochemical lipid determination, of fat mass in Caenorhabditis Elegans.[1]
Screw cap may be used for capping water samples in amber glass vial comprising of a polytetrafluoroethylene silicon septum for the analysis of chlorinated volatile organic compounds in water samples using headspace-solid-phase microextraction with gas chromatography (HS-SPME-GC).[2]
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Biochemical and high throughput microscopic assessment of fat mass in Caenorhabditis elegans.
Pino EC
Journal of Visualized Experiments, 73 (2013)
Determination of chlorinated volatile organic compounds in water and municipal wastewater using headspace-solid phase microextraction-gas chromatography.
Antoniou CV, et al.
Journal of Chromatography A, 1132(1-2), 310-314 (2006)