27088-U
Vials, screw top, amber glass (vial only)
volume 15 mL, amber glass vial, thread for 18-400, pkg of 100 ea
Synonym(s):
Amber Glass Screw Vials
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
amber glass vial
feature
closure type screw top vial
packaging
pkg of 100 ea
O.D. × H × I.D.
21 mm × 70 mm × 12 mm
volume
15 mL
fitting
thread for 18-400
Looking for similar products? Visit Product Comparison Guide
Related Categories
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service