Key Documents
25073
Supelco
Replacement septa for storage containers
greener alternative
for use with all TDS3 Containers
Select a Size
50 EA
€45.70
€45.70
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
50 EA
€45.70
About This Item
UNSPSC Code:
41104008
€45.70
Please contact Customer Service for Availability
Request a Bulk Order
packaging
pkg of 50 ea
greener alternative product characteristics
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
compatibility
for use with all TDS3 Containers
greener alternative category
General description
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
Simplify your thermal desorption-based terpenes sample collection in air. Learn more about their profiling, odor measurement, industrial hygiene, and environmental emissions.