Skip to Content
Merck
All Photos(1)

Key Documents

24916

Supelco

Purge/Trap G

configured for Tekmar® 3000, 3100 (Straight, 12 in./30.5 cm, Valco fitting

Synonym(s):

PURGE TRAP G, Tekmar® 3000

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€66.90

€66.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€66.90

About This Item

UNSPSC Code:
41115700

€66.90


Please contact Customer Service for Availability

Request a Bulk Order

application(s)

industrial hygiene

Quality Level

compatibility

configured for Tekmar® 3000, 3100 (Straight, 12 in./30.5 cm, Valco fitting

Looking for similar products? Visit Product Comparison Guide

General description

Tubes used in purge and trap analyses generally are packed with multiple beds of adsorbent materials, so that a broad range of polar and nonpolar, high and low molecular weight compounds can be trapped in a single tube. Each bed protects the next, increasingly active bed, by preventing compounds from being held so strongly that they cannot be desorbed quickly without decomposition. During the purge phase of sampling, lower molecular weight compounds pass through the initial adsorbent beds but are trapped by succeeding beds. During desorption, the carrier gas passes through the trap in the reverse direction of purge flow, so that higher molecular weight compounds never come in contact with the stronger (innermost) adsorbents.

Legal Information

Tekmar is a registered trademark of Tekmar Co.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Lungs, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service