Key Documents
24437
SGE Needles for RN syringes
volume 10 μL, N10-5, needle size 26 ga
Select a Size
5 EA
€61.50
€61.50
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
5 EA
€61.50
About This Item
UNSPSC Code:
41122001
€61.50
Please contact Customer Service for Availability
Request a Bulk Order
material
bevel tip stainless steel needle (point style 2)
needle size
26 ga
description
replacement needles
packaging
pkg of 5 ea
manufacturer/tradename
SGE 037110
volume
10 μL
General description
RN syringes allow easy replacement of needles that get damaged in the field. The needle is seated at zero graduation mark.
Choose from one of the most recent versions:
Certificates of Analysis (COA)