Key Documents
23632
Supelco
"Y" GlasSeal Connector, Fused Silica
pkg of 3 ea
Select a Size
3 EA
€341.00
€341.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
3 EA
€341.00
About This Item
UNSPSC Code:
41115700
€341.00
Please contact Customer Service for Availability
Request a Bulk Order
packaging
pkg of 3 ea
technique(s)
gas chromatography (GC): suitable
General description
Quickly connect three pieces of fused silica tubing of the same or different diameters without the need for tools. The leak-free connection is useful to split a sample to two columns, or to split a column effluent to two detectors. Silanized for an inert inside surface. For use with 0.10 - 0.53 mm I.D. fused silica tubing.
Application
"Y" GlasSeal Connector maybe used in capillary gas chromatographic-electroantennographic detection (GC-EAD) for chirality analysis of essential amino acid methyl esters.[1]
Other Notes
We offer a variety of chromatography accessories including analytical syringes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Essential amino acid methyl esters: major sex pheromone components of the cranberry white grub, Phyllophaga anxia (Coleoptera: Scarabaeidae).
Zhang A
Journal of Chemical Ecology, 23 (1), 231-245 (1997)