Key Documents
23318-U
Supelco
Gold-Plated Inlet Seal (Straight Design)
pkg of 2 ea
Select a Size
2 EA
€130.00
€130.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
2 EA
€130.00
About This Item
UNSPSC Code:
41121815
NACRES:
SB.54
€130.00
Please contact Customer Service for Availability
Request a Bulk Order
material
stainless steel (gold-plated)
packaging
pkg of 2 ea
General description
The inlet seal on an Agilent GC must be regularly changed to prevent sample adsorption due to accumulation of sample residue and/or septum fragments. Supelco manufactures replacement inlet seals using a high quality stainless steel plated with pure gold to insure inertness. No brighteners are used in the plating process. This straight design seal is suitable for applications with low split flows (<200 mL/min.). Each pack includes one washer for each seal.
Application
Gold-plated inlet seal may be used in the GC injection port for thermal desorption-gas chromatography/mass spectrometry (TD-GC/MS), where residual accumulation was noticed resulting in analyte adsorption and elevating signal background. Hence, replacing the gold-plated seal in regular intervals decreased the interference.[1]
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Precautions for in-injection port thermal desorption-gas chromatography/mass spectrometry (TD-GC/MS) as applied to aerosol filter samples.
Ho, Steven Sai Hang, et al.
Atmospheric Environment, 45, 1491-1496 (2011)
Articles
Pistachios 45% fat complicates pesticide analysis. But Z-Sep+ sorbent cleanup after QuEChERS extraction can reduce background for reliable results.
2-layer SPE cartridge efficiently reduces background for ginger contaminant analysis.
Assessment of clean-up sorbents for extracting PAHs from salmon and PCBs from milk, optimized for GC analysis with MS and ECD detection.
Cannabis testing regulations include analysis of purity, potency, and contaminants. Learn more about analysis of cannabis for pesticides, mycotoxins, and pyrethrins registered in California by LC-MS/MS and GC-MS/MS Using Ascentis® Express Biphenyl and SLB®-5ms Columns
Protocols
Paprika spice's PAH contamination risk due to environmental exposure highlights food safety concerns.