Skip to Content
Merck
All Photos(1)

Key Documents

22697

On/Off Toggle Valve

brass, 1/4 in. straight arms

Synonym(s):

Swagelok® 1GS4,BRASS TOGGLE VALVE 1/4X1/4S EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115717
eCl@ss:
32035302

material

brass

fitting

1/4 in. straight arms

Looking for similar products? Visit Product Comparison Guide

General description

Nuts and ferrules included.
Toggle valve on and off settings determines whether or not fluid flows past the valve. Well-designed toggle valve should possess the quality that it hinders flow completely when closed, while allowing nearly unaffected flow when open. Main goal of a toddle valve is to minimize the effect of pressure when in open position.[1]

Application

Toggle valve may be used in GC to describe a system which could eliminate the use of mechanical valves in the oven, and could be easily added to the existing chromatographs.[2]

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Angeli et al.
Molecules (Basel, Switzerland), 25(10) (2020-05-16)
Carbonic anhydrase (CA) is a zinc enzyme that catalyzes the reversible conversion of carbon dioxide to bicarbonate and proton. Currently, CA inhibitors are widely used as antiglaucoma, anticancer, and anti-obesity drugs and for the treatment of neurological disorders. Recently, the
Michael D. Holloway, Chikezie Nwaoha, Oliver A. Onyewuenyi
Process Plant Equipment: Operation, Control, and Reliability (2012)
A new technique for heart cutting in gas chromatography.
Deans, D. R.
Chromatographia, 1, 18-22 (1968)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service