Key Documents
22689
Swagelok® Port Connector
Swagelok® 201-PC, stainless steel, 1/8 in. Swagelok
Select a Size
2 EA
€48.70
€48.70
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
2 EA
€48.70
About This Item
UNSPSC Code:
41115717
€48.70
Please contact Customer Service for Availability
Request a Bulk Order
material
stainless steel
packaging
pkg of 2 ea
manufacturer/tradename
Swagelok® 201-PC
fitting
1/8 in. Swagelok
Application
Swagelok® port connector with two Swagelok® end caps may be used in construction of the reactors used in experimental studies investigating alkylarene pyrolysis coupled with mechanistic modeling.[1]
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Reactions of polycyclic alkylaromatics. 4. Hydrogenolysis mechanisms in 1-alkylpyrene pyrolysis
Smith, C. Michael, and Phillip E. Savage
Energy and Fuels, 6, 195-202 (1992)