Documents
22583
Supelco
Merlin Microseal System Replacement Septum
1 Low Pressure (1-45 psi) septum
About This Item
UNSPSC Code:
41115700
eCl@ss:
32049213
General description
The Merlin Microseal System is a septum-less system that provides very long life. Because there is not a septum to pierce, septum fragments will not be formed and deposited in the inlet liner. The septum contains a series of seals (wiper rib, o-rings, and a duckbill valve) that allow a needle to enter while maintaining a leak-free seal. The septum can be used only with a syringe that has a 23 gauge "blunt tipped" needle, or with an SPME fiber assembly with a 23 gauge needle. The thicker shaft is required to make the necessary contact with the septum seals.
Other Notes
Three septa versions are available:
  • Low Pressure for use with 23 gauge syringe needles, and injection port pressures between 1 and 45 psi. Do not use with syringe needles that have beveled tips.
  • General Purpose for use with 23 gauge syringe needles, and injection port pressures between 3 and 100 psi. Do not use with syringe needles that have beveled tips.
  • SPME for use with 23 gauge SPME fiber assemblies. Do not use with SPME fiber assemblies that have beveled tips.
Note: Do not use with beveled tips.
Legal Information
MicroSeal is a trademark of Merlin Instrument Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Determination of the plasticizer N-butylbenzenesulfonamide and the pharmaceutical Ibuprofen in wastewater using solid phase microextraction (SPME).
Huppert
Fresenius Journal of Analytical Chemistry, 362, 529-536 (1998)
J M Diserens
Journal of AOAC International, 84(3), 853-860 (2001-06-22)
Although the use of pentachlorophenol (PCP) is prohibited in most countries, it is still widely found in the wood of pallets, containers, crates and in cardboard, paper, etc. Such material may contain not only PCP but other chlorophenols as well.