Key Documents
22386
Supelco
Supeltex M-1 Packed Column Ferrule, 1/16 in. Column O.D.
I.D. 1/16 in., 1/16 in. O.D. Column, pkg of 10 ea
Select a Size
10 EA
€38.90
€38.90
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
10 EA
€38.90
About This Item
UNSPSC Code:
41121815
€38.90
Please contact Customer Service for Availability
Request a Bulk Order
packaging
pkg of 10 ea
I.D.
1/16 in.
fitting
1/16 in. ferrule for 1/16 in. O.D. packed column
compatibility
1/16 in. O.D. Column
General description
Characteristics: Ideal for connection to mass spectrometers. High reusability. Isothermal use only.
Material Composition: Ceramic-filled PTFE
Maximum Temperature:
250 °C
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Solid adsorbent preconcentration and gas chromatographic analysis of sulfur gases.
Black MS
Analytical Chemistry, 50 (7), 848-851 (1978)