Key Documents
22284
Push Button Valve
for use with VICI Series C and D Pressure-Lok syringes
Select a Size
1 EA
€30.90
€30.90
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
€30.90
About This Item
UNSPSC Code:
41122001
€30.90
Please contact Customer Service for Availability
Request a Bulk Order
material
PTFE Mininert valve
packaging
pkg of 1 ea
1 ea of
manufacturer/tradename
654050
VICI Precision Sampling
compatibility
for use with VICI Series C and D Pressure-Lok syringes
General description
This Mininert valve will attach to your syringe and provide sample storing capability. It features push-button operation and an all-PTFE body with a stainless steel miniature valve stem.
Choose from one of the most recent versions:
Certificates of Analysis (COA)