Key Documents
22024
Swagelok® Nut, Front and Back Ferrule Set
Swagelok® 100-S, brass, 1/16 in. Swagelok
Select a Size
10 PKG
€69.60
€69.60
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
10 PKG
€69.60
About This Item
UNSPSC Code:
41115717
€69.60
Please contact Customer Service for Availability
Request a Bulk Order
material
brass
packaging
pkg of 10 ea
manufacturer/tradename
Swagelok® 100-S
fitting
1/16 in. Swagelok
General description
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut.[1] While assembling a compression tube fitting, the metal tubing should be inspected properly to ensure that the surface is smooth. Then slide on the nut with the open side facing the end of the tubing. Next, slide on the back ferrule with the wider part facing the nut. In the next step, slide on the front ferrule with the small end of the cone facing the end of the tubing. Then insert the tubing into the fitting and tighten up the nut/ferrule assembly onto the fitting.[2]
Application
Swagelok® nut, front and back ferrule set maybe used for sampler construction from a single capillary gas chromatographic column, for collection of semi-volatile organic compounds through a diffusion denuder.[3]
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Diffusion denuder for the collection of semivolatile organic compounds.
Krieger MS and Hites RA.
Environmental Science & Technology, 26 (8), 1551-1555 (1992)
Grob RL & Barry EF.
Modern Practice of Gas Chromatography, 522-523 (2004)
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)