21740-U
Precision Sampling syringe needles
volume 50-500 mL, for use with VICI Magnum Series, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2.0 in. × 0.035 in. × 0.023 in.
Synonym(s):
REPLACEMENT TAPER NEEDLE 20 GA X 2" 3/PK
About This Item
Recommended Products
material
bevel tip stainless steel needle (point style 2)
description
Magnum Series
packaging
pkg of 3 ea
manufacturer/tradename
VICI 953050
needle L × O.D. × I.D.
2.0 in. × 0.035 in. × 0.023 in.
volume
50-500 mL
compatibility
for use with VICI Magnum Series
Looking for similar products? Visit Product Comparison Guide
General description
- Ideal for collection, trace analysis and sample concentration of environmental contaminants.
- A low-dead volume needle is used in this syringe.
- A rotary valve allows on-site sampling for later analysis.
- Positive displacement capability of the durable PTFE plunger seal permits use in a variety of subsequent procedures involving extraction or concentration of trace components.
The standard termination is a female threaded fitting (1/4-28), which accepts a variety of Cheminert connectors or adapters. Specially formed seals permit leak-tight operation to 30 psi (approximately 2 atmospheres).
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service