Skip to Content
Merck
All Photos(1)

Documents

21284-U

Supelco

GC Stationary Phase

phase SP®-2100, bottle of 10 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
SB.54

Agency

meets requirements for USP G1

packaging

bottle of 10 g

parameter

0-350 °C temp. range

technique(s)

gas chromatography (GC): suitable

matrix active group

SP®-2100 phase

solubility

chloroform: soluble

column type

packed GC

General description

GC methods are divided into two classes depending on the nature of stationary phases; gas-solid chromatography (GSC) and gas-liquid chromatography (GLC). GSC has solid adsorptive material and solute particles are removed from mobile phase by electrostatic forces. GLC has a thin layer of liquid coated or bonded on the surface of an inert particle or on the walls of the column where solute particles are retained in the liquid phase based on their partition coefficients. The primary necessity of a stationary phase is to provide sample separation sustaining phase integrity over a reasonable period of time. It should be stable for the chemical and thermal changes. Selectivity, peak symmetry, analysis time, degree of separation, peak tailing are a few parameters that should be considered in order to choose a stationary phase. SP-2100 is a methyl silicone fluid suitable as stationary phase.
Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.

Application

SP-2100 fused silica column deactivated with Carbowax 20M is suitable for determining optimal conditions for the analysis of fatty and resin acids in wood resin.

Legal Information

SP is a registered trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

214.0 °F - closed cup

Flash Point(C)

101.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Operating variables in the analysis of tall oil acids by capillary gas chromatography.
Dorris, G. M.
Journal of the American Oil Chemists' Society, 59, 494-500 (1982)
Stanley Blackburn
Peptides, 286-286 (1986)
David B. Troy, Paul Beringer
Remington: The Science and Practice of Pharmacy, 605-605 (2006)
James P. Lodge, Jr.
Methods of Air Sampling and Analysis, 98-98 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service