21233-U
ORBO™ 1500 Precleaned Small PUF/Amberlite™ XAD®-2/PUF Cartridge
O.D. × L 22 mm × 10 cm, pkg of 3 ea
Synonym(s):
Small PUF Cartridges
About This Item
Recommended Products
material
polyurethane (foam)
description
PUF/XAD-2/PUF Layers; Each PUF 30 mm L; 1.5g Amberlite™ XAD™-2
assembled
product line
ORBO™
feature
precleaned
packaging
pkg of 3 ea
manufacturer/tradename
ORBO™ 1500
technique(s)
active air sampling: suitable
O.D. × L
22 mm × 10 cm
suitability
suitable for (Research pesticides)
suitable for (Semivolatiles (pesticides, PCBs, PAHs))
application(s)
air monitoring
environmental
industrial hygiene
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service