Key Documents
2049801
Supelco
Inlet liner, split with cup (unpacked) for Varian® 1075/1077 injector
About This Item
UNSPSC Code:
41121800
Pricing and availability is not currently available.
material
glass
description
Inlet Liner, Split with cup, for Varian
Inlet liner
cup yes unpacked
feature
deactivation high temperature silanization
packaging
pkg of 1 ea
manufacturer/tradename
Varian 01-900109-02 (OEM)
L × O.D. × I.D.
72 mm × 6.3 mm × 4 mm
liner L × O.D.
72 mm × 6.3 mm
compatibility
configured for
for use with Varian 1075/1077 Injectors
for use with Varian
technique used for High & low MW analytes
Legal Information
Varian is a registered trademark of Varian Medical Systems, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)