Skip to Content
Merck
All Photos(1)

Key Documents

18542

Supelco

Cyclohexanol

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

18542-100MG
€128.00

€128.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
18542-100MG
€128.00

About This Item

Linear Formula:
C6H11OH
CAS Number:
Molecular Weight:
100.16
Beilstein:
906744
MDL number:
UNSPSC Code:
12352200

€128.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

vapor density

3.5 (vs air)

vapor pressure

0.98 mmHg ( 25 °C)

autoignition temp.

572 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

12.25 %

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

refractive index

n20/D 1.465 (lit.)

bp

160-161 °C (lit.)

mp

20-22 °C (lit.)

density

0.948 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OC1CCCCC1

InChI

1S/C6H12O/c7-6-4-2-1-3-5-6/h6-7H,1-5H2

InChI key

HPXRVTGHNJAIIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup - DIN 51794

Flash Point(C)

64 °C - closed cup - DIN 51794


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service